Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
HPLC columns, NUCLEOSIL® 300 C18
Supplier: Hichrom Limited
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
NC300-5C18-10CM5EA 0 INR InStock
HICHNC300-5C1850AS HICHNC300-5C1850AM HICHNC3005C1810CM5 HICHNC300-5C18250A HICHNC3005C18250A HICHNC300-5C18150A HICHNC300-5C18100A HICHNC300-5C1810C5
HPLC columns, NUCLEOSIL® 300 C18
These NUCLEOSIL® HPLC columns are packed with spherical porous silica available in a pore size of 300 Å. This silica is used as the base material for reversed phase bioanalytical columns.
  • Wide range of bonded phases
  • Good mechanical stability
  • Highly efficient
Surface area: 100 m²/g
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms