HPLC columns, NUCLEOSIL® 100 C18 AB
Supplier: Hichrom Limited
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Nucleosil®
NC1005C18B50AFEA 0 INR
HICHNC1005C18B50AF HICHNC1005C-18-150 HICHNC1005C18AB50A HICHNC1005C18AB100 HICHNC1005C-18-250 HICHNC1005C18B10C5 HICHNC1005C18AB150 HICHNC1005C-18-100 HICHNC1005C18B250A HICHNC1005C18B150A
HPLC columns, NUCLEOSIL® 100 C18 AB
Chromatography Columns
Packed with classical spherical porous silica available in a wide range of pore sizes. The 100 Å and 120 Å have similar relative retention characteristics, however the 100 Å has the greater capacity.
  • Good mechanical stability
  • Good batch to batch reproducibility