1-Pentanesulphonic acid sodium salt for HPLC, Rankem®

Supplier: RANKEM (A AVANTOR BRAND)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Sodium pentanesulphonate, 98%Sodium pentane-1-sulphonatePentane-1-sulphonic acid sodium saltSodium pentanesulphonateSodium 1-pentanesulphonate

RANKP2109EA 2125.2 INR
RANKP2109
1-Pentanesulphonic acid sodium salt for HPLC, Rankem®
1-Pentanesulphonic acid sodium salt
Formula: C₅H₁₁O₃SNa
MW: 174,2 g/mol
Melting Pt: Min. 300 °C
Storage Temperature: ambient
MDL Number: MFCD00007541
CAS Number: 22767-49-3
EINECS: 245-208-4
UN: 0000
https://b2b-in-gqp.vwr.com/stibo/web/std.lang.all/09/27/18570927.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR