ProClean Plus, J.T.Baker®

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


J.T.Baker®
3901.EA 0 INR
BAKR3901
ProClean Plus, J.T.Baker®
Performance Test: passes test
pH at 25 °C: 8.5 - 9.1
Total Bacterial Growth (CFU/100ml): max. 50
Absorbance maximum at 595 nm: 1,40 - 1,80

Caution: pH is measured directly after production. In course of time the actual value will decrease. This will not influence the performance of the reagent.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR