HPLC columns, NUCLEOSIL® 100 10CN
Supplier: Hichrom Limited
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Nucleosil®
NC10010CN100AEA 0 INR
HICHNC10010CN100A HICHNC10010CN10C5 HICHNC10010CN250AF HICHNC10010CN150A HICHNC10010CN150AF HICHNC10010CN300AF HICHNC10010CN200AN HICHNC10010CN200A
HPLC columns, NUCLEOSIL® 100 10CN
Chromatography Columns
Packed with classical spherical porous silica available in a wide range of pore sizes. The 100 and 120 Å have similar relative retention characteristics, however 120 Å exhibits a lower backpressure.
  • Good mechanical stability
Caution: *Use with freestanding holder HI-161 and column coupler HI-081, please see 'HPLC Guard Column Holders and Accessories'