Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

BeneSphera™ HbsAg ELISA Kit

Supplier: BENESPHERA

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:


15001007 15001548 15001549
BENP15001007EA 1874.17 INR InStock
BENP15001007 BENP15001548 BENP15001549
BeneSphera™ HbsAg ELISA Kit
Assays ELISAs
BeneSphera™ HbsAg ELISA kit is for the qualitative detection of Hepatitis B Surface Antigen from human serum and plasma.

  • Sandwich, non-competitive, immunoassay with HRP as a marker enzyme
  • Detects all major 10 serotypes
  • Analytical Sensitivity: 0,2 IU/ml
  • Diagnostic sensitivity: 100%
  • Diagnostic specificity: 99,5%

BeneSphera™ HBsAg Microwell ELISA (IHF) kit represents a new generation of HBsAg assays in which specific monoclonal Anti-HBs antibodies are used. This assay promises to be more sensitive, convenient and safer to use, and has advantage of short reaction time as well as easier detection of target materials.

Delivery information: Kit includes: Monoclonal anti-HBs coated microstrips; Control (positive/negative); Conjugate concentrate: 50X; Conjugate diluent; TMB chromogen in DMSO; Substrate buffer; Wash solution; Stop solution; Empty zip seal polybag; Plate sealer in zip seal polybag.

Caution: The material is for invitro diagnostic use only.


Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms