Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Ultra high temperature seal ND20 with central hole, VWR®
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
548-2378EA 109666 INR InStock
VWRI548-2378
Ultra high temperature seal ND20 with central hole, VWR®
High temperature, high quality Silicone/PTFE septum for less extractables at low to mid-high temperatures.
  • Operation upto 300 °C possible.
  • Low bleeding level at high temperatures less than 120 °C
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms