EDTA disodium salt dihydrate AR, Rankem®
Supplier: RANKEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E0127
E0120
E0122
RANKE0127EA
12069.75
INR
RANKE0127
RANKE0120
RANKE0122
EDTA disodium salt dihydrate AR, Rankem®
EDTA disodium salt dihydrate
Formula:
C₁₀H₁₄N₂Na₂O₈·2H₂O MW: 372.24 g/mol Melting Pt: 255 |
MDL Number:
MFCD00150037 CAS Number: 6381-92-6 Merck Index: 13,03543 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...