Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Acrylogel 2.6 solution 30%, Electran for electrophoresis, VWR®
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
427205EEA 0 INR InStock
VWRC427205E
Acrylogel 2.6 solution 30%, Electran for electrophoresis, VWR®
A convenient, ready to use 30% w/v (30%T) solution of acrylamide `Electran` and NN'-methylenebisacrylamide `Electran` (2.6% C) in deionised water.
(Final ratio is 37.5:1)
Order Now
ORDER
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms