Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Polyvinyl alcohol, polymer with vinyl acetate, 25/140, RHODOVIOL®

Supplier: VWR Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:


20954.295EA 5056 INR InStock
20954.295
Polyvinyl alcohol, polymer with vinyl acetate, 25/140, RHODOVIOL®
Polyvinyl alcohol, polymer with vinyl acetate, 25/140


CAS Number: 25213-24-5
EINECS: 000-000-0
UN: 0000
Order Now
Specification Test Results
Appearance Granular powder
Solvent Water
Acidity Max. 0.08 meq/g
Apparent density 0.530 - 0.600
Ester value 120 - 150
pH (20°C) 5 - 7
Viscosity (20°C; 4 %; water) 24 - 30 mPa.s
Ignition residue (as Na2O) Max. 0.5 %
Volatile matter Max. 5 %

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms