γ-(N-Morpholino)propanesulphonic acid (MOPS) ≥99%, Ultrapure

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-(N-Morpholino)propanesulphonic acid3-Morpholinopropane-1-sulphonic acid

0670-500GEA 10247 INR
0670-500G 0670-250G
γ-(N-Morpholino)propanesulphonic acid (MOPS) ≥99%, Ultrapure
γ-(N-Morpholino)propanesulphonic acid (MOPS)

MOPS is a zwitterionic buffer used as a running buffer for denaturing agarose gel electrophoresis of RNA.

Formula: C₇H₁₅NO₄S
MW: 209,27 g/mol
Melting Pt: >250 °C
Flash Pt: 110 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5
UN: 0000
Merck Index: 13,06288

Order Now

Specification Test Results

Abs.@260 nm (0.1 M, Water) ≤0.05
Abs.@280 nm (0.1 M, Water) ≤0.05
DNase none detected
RNase none detected
pKa@(25 °C) 7 – 7.4
Protease none
Purity >99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR