VWRC44383
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
443832TEA 0 INR
VWRC443832T
VWRC44383
MOPS is a zwitterionic amino acid which acts as one of the "Good" buffers. It is a structural analog to MES. However, MOPS is a more suitable biological buffer than MES because the pH range is closer to pH 7.4.