Methylene blue
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Basic Blue 9
, Swiss Blue
, Methylthioninium chloride
, Methylene blue chloride
, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride
A1402.0100EA
0
INR
A1402.0100
Methylene blue
Methylene blue
Formula:
C₁₆H₁₈ClN₃S MW: 319.86 g/mol Melting Pt: 180 °C Density: 1.757 g/cm³ (22 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150008 CAS Number: 61-73-4 EINECS: 200-515-2 UN: 0000 Merck Index: 12,06137 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...