Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

L(+)-Glutamine kit 98.5-101.5% (dry basis) USP

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:


2078-K1EA 107787.06 INR InStock
2078-K1 2078-K2
L(+)-Glutamine kit 98.5-101.5% (dry basis) USP
L(+)-Glutamine


Formula: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 185…186 °C
Density: 1.5326 g/cm³ (130 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008044
CAS Number: 56-85-9
EINECS: 200-292-1
Merck Index: 13,04484
Order Now
Specification Test Results
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
GMP Manufactured Product
Bulk Pharmaceutical Chemical
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Kit Components Meet Requirements Passes Test
This kit contains one each of L-Glutamine 2078-B1 and 2078-SA.

 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms