Polyvinyl alcohol 87-89%, partially hydrolysed, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIALS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Mowiol®
, Poly(vinyl alcohol)
, PVOH
, PVA
Management of Change (MOC) category = N
U23208EA
42398.8
INR
U23208
Polyvinyl alcohol 87-89%, partially hydrolysed, J.T.Baker®
Polyvinyl alcohol
Formula:
[-CH₂CH(OH)-]n |
MDL Number:
MFCD00081922 CAS Number: 9002-89-5 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Viscosity of 4% Aqueous Solution at 20°C (cps) | 4 - 7 |
Identification (by IR) | Passes Test |
pH of 4% Aqueous Solution | 4.5 - 6.5 |
Loss on Drying at 105°C | ≤5 % |
Residue after Ignition | ≤0.75 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...