Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

L(+)-Glutamine kit 98.5-101.5% (dry basis) USP, Multi-Compendial

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:


Management of Change (MOC) category = R

2078-B1EA 46146.55 INR InStock
2078-B1 2078-B2 2078-SA
L(+)-Glutamine kit 98.5-101.5% (dry basis) USP, Multi-Compendial
L(+)-Glutamine


Formula: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 185…186 °C
Density: 1.5326 g/cm³ (130 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008044
CAS Number: 56-85-9
EINECS: 200-292-1
Merck Index: 13,04484
Order Now
Specification Test Results
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₅H₁₀N₂O₃) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Specific Rotation [Å]²º^D (+) 6.3 - 7.3 Degree
USP - Loss on Drying at 105°C ≤ 0.3 %
USP - Residue after Ignition ≤ 0.3 %
USP - Chloride (Cl) ≤ 0.05 %
USP - Sulfate (SO₄) ≤ 0.03 %
USP - Iron (Fe) ≤ 0.003 %
USP - Related Compounds 0.5% max
JP - Assay (C₅H₁₀N₂O₃) (dried basis) 99.0 - 101.0 %
JP - Identification Passes Test
JP - Specific Rotation [Å]²º^D (+) 6.3 - 7.3 Degree
JP - Loss on Drying at 105°C ≤ 0.30 %
JP - Residue after Ignition ≤ 0.1 %
JP - Chloride (Cl) ≤ 0.021 %
JP - Sulfate (SO₄) ≤ 0.028 %
JP - Iron (Fe) ≤ 10 ppm
JP - Heavy Metals (as Pb) ≤ 10 ppm
JP - Related substances Passes Test
JP - pH 4.5 - 6.0
JP - Clarity and Color of Solution Passes Test
JP - Ammonium ≤ 0.1 %
Solubility of 3% Solution at 30°C Passes Test
Solubility of 3.5% Solution at 30°C Passes Test
Identification (by IR) Passes Test
Endotoxin Concentration (EU/mg) ≤ 0.06
Endotoxin Concentration, IU/mg Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.
This product has been filled to +/- 1% of the stated net weight.

 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms