Urea 99.0-100.5% USP, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Carbonyl diamideCarbamideCarbonyldiamideDiaminomethanone

Management of Change (MOC) category = R

4205-60EA 595065.38 INR
4205-60
Urea 99.0-100.5% USP, J.T.Baker®
Urea
Formula: NH₂CONH₂
MW: 60,06 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 132.7 °C
Density: 1,34 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
UN: 0000
REACH: 01-2119463277-33
Merck Index: 13,09935

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (NH₂CONH₂) 98.0 - 102.0 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Residue on Ignition ≤0.1 %
USP - Alcohol Insoluble Matter ≤0.04 %
USP - Organic impurities: Urea related compound A ≤0.1 %
USP - Organic impurities: Any individual unspecified impurity ≤0.1 %
USP - Organic impurities: Total impurities ≤2.0 %
Solution Test Passes Test
Non-Metallic Impurities - Arsenic (As), For Information Only
Metallic Impurities - Aluminum (Al), For Information Only
Metallic Impurities - Barium (Ba), For Information Only
Metallic Impurities - Bismuth (Bi), For Information Only
Metallic Impurities - Cadmium (Cd), For Information Only
Metallic Impurities - Calcium (Ca), For Information Only
Metallic Impurities - Chromium (Cr), For Information Only
Metallic Impurities - Cobalt (Co), For Information Only
Metallic Impurities - Lead (Pb), For Information Only
Metallic Impurities - Lithium (Li), For Information Only
Metallic Impurities - Magnesium (Mg), For Information Only
Metallic Impurities - Manganese (Mn), For Information Only
Metallic Impurities - Mercury (Hg), For Information Only
Metallic Impurities - Molybdenum (Mo), For Information Only
Metallic Impurities - Nickel (Ni), For Information Only
Metallic Impurities - Potassium (K), For Information Only
Metallic Impurities - Sodium (Na), For Information Only
Metallic Impurities - Strontium (Sr), For Information Only
Metallic Impurities - Vanadium (V), For Information Only
Metallic Impurities - Zinc (Zn), For Information Only
Filtered through a 0.2 micron filter.
Residual Solvents: No Class 1,2,3 or other solvents are used or produced
in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR