Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Safety goggles with face shield, KleenGuard™ V90 shield
Supplier: Kimberly-Clark
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
KleenGuard™
KIMB18629EA 0 INR InStock
KIMB18629
Safety goggles with face shield, KleenGuard™ V90 shield
Goggles
KleenGuard™ Personal Protective Equipment (PPE) enables rather than inhibits – allowing for maximum productivity and safety. They provide eye and full face protection from liquids and particles without the hassle of primary eye protection underneath a face shield mask.
  • Monogoggle XTR goggles, so there is no need to wear primary eye protection, like with other face shields
  • Often fits over most prescription eyeglasses
  • Feature an indirect vent that enables free airflow, while safeguarding against liquids entering the eye area
  • Polycarbonate lenses provide 99,9% UVA / UVB / UVC protection
  • Shield has a D3 rating for droplet and splash protection
  • Clear anti-fog lens helps workers see clearly, even in humid or wet environments
The goggles portion has a soft, pliable frame that conforms to the face for comfort, a cylindrical lens for increased visibility, and indirect ventilation. They can fit over most prescription eyeglasses. The face shield mask is curved to conform to the shape of face for added protection. The Shield includes an adjustable strap, so that your workers can get a snug fit.
Certifications: Meets ANSI Z87.1+ standard for impact protection, with a D3 rating for droplet and splash protection.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms