VWR®, Micro-centrifuge, Benchtop, High Speed
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWRA460-0365EA 31050 INR
VWRA460-0365 VWRA521-3307
VWR®, Micro-centrifuge, Benchtop, High Speed
Centrifuges Microcentrifuges
Delivering accuracy in every cycle even with heavy loads, this high speed benchtop micro-centrifuge has been designed with the users' requirements in mind. The lightning fast 15000 min⁻¹ microprocessor controlled centrifuge can perform at peak efficiency and give the desired results, it gives smooth and silent operation even at maximum speed.
  • Microprocessor controlled efficient performance
  • Variable speed from 500 to 15000 min⁻¹
  • Intuitive interface for quick and convenient setting with last run memory feature
  • Smooth and silent operation even at maximum speed
  • Large LCD display enables multiple parameters to be viewed at a single glance
  • One-touch short spin
  • One touch RPM / RCF conversion operation
  • Safe low voltage device
Supplied with choice of two types of rotor, please order as per requirements of the application.
Ordering information: Rotors supplied as standard accessory, please choose the model as per rotor requirements.