Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Hydrochloric acid 0.25 N, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Warning
Synonyms: Muriatic acidHydrochloric acid solution

Management of Change (MOC) category = R

0322-09EA 0 INR InStock
0322-09
Hydrochloric acid 0.25 N, Biotech Reagent, J.T.Baker®
Hydrochloric acid


Formula: HCl
MW: 36,46 g/mol
Storage Temperature: ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III
Order Now
Specification Test Results
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Ammonium (NH₄) ≤ 3 ppm
Appearance (Clear, colorless liquid) Passes Test
Trace Impurities - Heavy Metals ≤ 0.5 ppm
Normality, Eq/L 0.23 - 0.27
Sulfate (SO₄) ≤ 1 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Endotoxin concentration, EU/mL ≤ 1

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms