Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Ammonium peroxodisulphate (APS), Microbiological Grade

Supplier: VWR Collection

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger
Synonyms: Peroxidisulphuric acid diammonium saltAmmonium persulphate

VWR®
VWRA06190110EA 5200 INR InStock
VWRA06190110
Ammonium peroxodisulphate (APS), Microbiological Grade
Ammonium peroxodisulphate (APS)
VWR Delivered High quality Reagents & biochemicals, combined with innovative products that gives optimum results , save time & simplify protocols

  • VWR Lifescience Molecular Biology grade Reagents & Biochemicals.


Formula: (NH₄)₂S₂O₈
MW: 228,2 g/mol
Melting Pt: 120 °C
Density: 1,98 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00003390
CAS Number: 7727-54-0
EINECS: 231-786-5
ADR: 5.1,III
Merck Index: 13,00545
Order Now

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms