Pyrex Round-Bottom Centrifuge Tubes, Glass
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX34533-50EA 0 INR
MFLX34533-50 MFLX34533-01
Pyrex Round-Bottom Centrifuge Tubes, Glass
Tubes Centrifuge Tubes
Round-bottom tubes are available in rimless and screw-cap styles. Rimless tubes have heavy, fire-polished ends. Caps resist heat and steam. Corex®II and Pyrex® brand centrifuge tubes are made from durable Type I borosilicate glass. Corex®II is chemically strengthened for higher speed centrifugation.
PYREX® Corning Glass Works.