Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Thermo Scientific Nunc Round-Bottom Tubes
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
06329-31 06329-55 06329-60 06329-62 06330-31 06330-32 06330-40 06330-55 06330-60 06330-70 06330-75 06329-72
MFLX06329-31EA 0 INR InStock
MFLX06329-31 MFLX06329-55 MFLX06329-60 MFLX06329-62 MFLX06330-31 MFLX06330-32 MFLX06330-40 MFLX06330-55 MFLX06330-60 MFLX06330-70 MFLX06330-75 MFLX06329-72
Thermo Scientific Nunc Round-Bottom Tubes
Tubes Centrifuge Tubes
Withstands RCF up to 30,000 ×g.
  • Autoclavable
These PTFE round-botom tubes withstand relative centrifugal force (RCF) up to 30000 ×g in a close fitting holder. They are available with or without screw caps. Autoclavable. All other tubes withstand RCF up to 50,000 ×g when used in a refrigerated centrifuge. All tubes without caps accept snap-on polypropylene closures.
Thermo Scientific Nalgene® Nalge Thermo Scientific Nunc International
TEFZEL® E.I. DuPont de Nemours & Co.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms