Thermo Scientific Nalgene Polypropylene Sealing Caps
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
06327-65 06327-69
MFLX06327-65EA 0 INR
MFLX06327-65 MFLX06327-69
Thermo Scientific Nalgene Polypropylene Sealing Caps
Closures Bottle Closures
Ensures leakproof performance.
  • Autoclavable
These sealing caps are designed for use with Oak Ridge tubes and centrifuge bottles. Recommended for use when spinning hazardous materials. Retained silicone sealing ring requires no assembly.
Warranty: 1 year