Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
ETFE Luer Adapters
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX06471-85EA 0 INR InStock
MFLX06471-85
ETFE Luer Adapters
Fittings and Connectors
ETFE (or Tefzel®) adapters connect standard microbore tubing to luer fitting systems and are designed for use with low-pressure range chromatography fittings. Adapters with caps include zero dead volume PTFE cones and screw into the fitting body directly. Adapters with 1/4-28 UNF thread come with lock nut and screw directly into existing ports (does not include part number 06471-85).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms