Masterflex® Direct-Reading Variable-Area Flowmeters, Dual-Scale, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Masterflex®
MFLX03230-10EA 0 INR
MFLX03230-10
Masterflex® Direct-Reading Variable-Area Flowmeters, Dual-Scale, Avantor®
Flow Meters
Design makes each meter suitable for a broad range of laboratory applications.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium no need to convert readings to get flow rate
  • Dual-unit scales for air or water offer flexibility
Choose from unshielded, shielded, or shielded with valve flow meters suitable for a wide range of applications. Unshielded Flowmeters work well in low-pressure applications. Connect the flowtube end directly to tubing having the proper inner diameter. Maximum pressure rating for unshielded flowmeters is 15 psi. Shielded Flowmeters are better suited to higher-pressure applications or for installations requiring a panel-mounted flowmeter. End bushings are molded with dual connection capability-tubing or threaded. Shielded Flowmeters with Valves offer flow monitoring along with precise control through an integrated 20-turn micrometer valve. The valve can be adjusted from 0,1 to 100% of maximum flow (semilogarithmic) and provides recise regulation from 0,3 to 60% of maximum flow.
Glass taper joints are available upon request please contact your local Avantor office to order.