GF Signet Galvanized Iron (Schedule 80) Saddle Installation Fittings
Supplier: GF Signet
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX05620-58EA 52298.42 INR
MFLX05620-58
GF Signet Galvanized Iron (Schedule 80) Saddle Installation Fittings
Flow Meters
Ensure proper insertion depth and measurement.
  • Constructed of durable Galvanized Iron (80)
These GF Signet Galvanized Iron (80) tee installation fittings feature a saddle connection.