GF Signet Installation Fitting, Polypropylene Dummy Plug
Supplier: GF Signet
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX05614-29EA 16296.4 INR
MFLX05614-29
GF Signet Installation Fitting, Polypropylene Dummy Plug
Flow Meters
Quick and easy assembly without tools.
  • Constructed of durable polypropylene
Dummy Plugs -- Insert a plug in place of the sensor when it has been removed for inspection or service. Note: Not for use with METALEX sensors.