Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Mini Submersible Magnetic Drive Pumps
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
07105-05 07142-02 07142-15 07142-23 07142-17 07142-31
MFLX07105-05EA 52800 INR InStock
MFLX07105-05 MFLX07142-02 MFLX07142-15 MFLX07142-23 MFLX07142-17 MFLX07142-31
Mini Submersible Magnetic Drive Pumps
Pumps Centrifugal Pumps
Operate fully submerged or in open air.
  • Liquid-cooled models keep heat-sensitive fluids from warming up
  • Thick (1/8") epoxy coating dissipates heat quickly and provides good chemical resistance
  • UL-Listed
These mini submersible magnetic drive pumps are good for low-viscosity liquids including solvents, dyes, inks, and mild corrosives.
Delivery information: Includes a three-wire cord.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms