Flux® Heavy-Duty Drum Pump Motors, TEFC
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
07076-20 07076-25 70603-60
MFLX07076-20EA 323000 INR
MFLX07076-20 MFLX07076-25 MFLX70603-60
Flux® Heavy-Duty Drum Pump Motors, TEFC
Pumps Drum/Transfer Pumps
Configurable to your requirements.
  • High-viscosity tube set handles fluids with viscosities to 50,000 cp
Heavy-duty motors are best for applications requiring high output head. Motors have a built-in overload switch with manual rest to avoid the danger of automatic restarting.
Order tube set and accessories separately.