Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Masterflex® Ismatec® Compact Integrated Drive for Micropump® Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Masterflex®
MFLA78008-15EA 0 INR InStock
MFLA78008-15 MFLA78004-47 MFLX78004-37
Masterflex® Ismatec® Compact Integrated Drive for Micropump® Pump Heads, Avantor®
Pumps Pump Drives Gear Pump Drives
High-performance drives with numerous features.
  • Control speed and start/stop remotely via analog signal; RS-232 available on Reglo Digital and MCP-series drives
  • Magnetically driven pump heads provide smooth, accurate, pulseless fluid delivery
  • Don't forget your pump head and tubing for a complete pump system
  • Adapter plate to accept either A or B Mount gear pump heads is included
Compatible with all Micropump® A-mount (with adapter plate 07617-90 - included) or B-mount pump heads. Country specific power cord shipped with drive - chosen based upon ship-to address.
Two main gear pump styles fit the drives (pump heads sold separately):
Cavity style pump head can be used for viscous media and for applications with a certain suction lift, has smooth operation at a low noise, and low inner friction.
Suction shoe pump head features enhanced pump performance at elevated differential pressures and has fewer parts for easy maintenance. These pump heads are suitable for elevated temperature ranges and not suitable for applications requiring a suction lift.
Some gear pump head models have an internal fluid bypass. A fluid bypass valve helps protect against decoupling and damage from high pressure build-up.
Delivery information: Includes an adapter plate for mounting Micropump® A-mount pump heads, a driving magnet, and a 6-ft power cord with U.S. standard plug. Modular drives also include a 6-ft cable for connecting drive and controller. These models are shipped with country specific cords.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms