Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Poly-Cone™ Standard Taper Adapter, Polypropylene
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX06432-00EA 4800 INR InStock
MFLX06432-00
Poly-Cone™ Standard Taper Adapter, Polypropylene
Adapters Adapters, Conical Ground Joints
Forms a continuous standard taper inside and out, from top to bottom.
  • Cut into various adapters, such as individual bushings, stirrer bearings, thermometer and gas inlet tube supports, drip-tips, funnels, trap connectors for tubing, etc.
Polypropylene cone is 27-cm (10-5/8") long; cut to fit your needs. Tapers from 30- to 3-mm ID (35- to 8-mm OD). Withstands temperatures to 250ºF (121ºC). Bel-Art.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms