Gilmont Capillary Valves
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX03237-33EA 0 INR
MFLX03237-33 MFLX03235-00
Gilmont Capillary Valves
Valves Needle Valves
Attain extremely precise, reproducible flow control of both liquids and gases. Valves have a precision-bore Pyrex® glass capillary tube, PTFE rod and stem, and Viton® O-ring. Achieve semilogarithmic flow control in the first 97% of travel with micrometer valve; 70% with metering valve.
Micrometer valve includes Viton O-rings for 10–6 mm Hg vacuums and PTFE O-rings for 10–2 mm Hg vacuums. Simple-design metering valve works with high vacuums.
Quick-opening metering valve allows fast opening and closing with a small degree of flow regulation. All valves withstand maximum pressures of 30 psi and maximum temperatures of 250 °F (121 °C).