Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Hayward® True Union Ball Check Valves with Foot Valve Screens
Supplier: Hayward
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX01341-52EA 11772.73 INR InStock
MFLX01341-52 MFLX01341-58 MFLX01341-50
Hayward® True Union Ball Check Valves with Foot Valve Screens
Valves Check Valves
Unique square-cut elastomer seat provides seal at very low back pressures.
  • Suitable for use with Hayward® True Union Ball Check Valves
Quickly and easily install either vertically or horizontally. These valves are assembled with water soluble, silicone-free lubricant and constructed of a one-piece PVC body.
The foot valve screen helps prevent unwanted material from entering the system.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms