Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Burkert Two-Way Valves for Steam (Normally Closed)
Supplier: Burkert
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX98621-00EA 36294.59 INR InStock
MFLX98621-00
Burkert Two-Way Valves for Steam (Normally Closed)
Valves Solenoid Valves
Fast-acting long life solenoid valves.
  • Constructed with brass body and PTFE seals
Two-Way Valves for Steam have a brass body and PTFE seals. The high temperature range of 32 to 356 °F (0 to 180 °C) makes these valves ideal for steam, hot water, oils, and other fluids compatible with the wetted materials.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms