Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Flux® Drum Pump Fume Glands
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
07076-62 07076-72
MFLX07076-62EA 88100 INR InStock
MFLX07076-62 MFLX07076-72
Flux® Drum Pump Fume Glands
Pumps Drum/Transfer Pumps
Protect your drum pump.
  • Choose the material and size suitable for your application
Fume Glands do not allow fumes from the liquid in the drum to vent out. Gland allows air to enter the drum when the pump is operating, then closes when the pump is switched off and left in position in the drum.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms