Flux® Drum Pump Mixing Tube Sets
Supplier: Flux Pumps
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX07076-50EA 311453.01 INR
MFLX07076-50 MFLX07076-54
Flux® Drum Pump Mixing Tube Sets
Pumps Drum/Transfer Pumps
Configurable drum pump components to fit your requirements.
  • Choose between stainless steel (SS) or polypropylene (PP) construction
These new tube sets eliminate the hassle of needing a separate mixing tube. With the unique lever design, you can easily switch between pumping and mixing modes to pump and mix the drum contents separately or simultaneously. SS tube sets are 19/16" OD with 1" hose barb; PP tube sets are 2" OD with 3/4", 1", or 1-1/4" hose barb.