Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Iron Reduction Cartridge for "Big Blue" housing
Supplier: Pentair
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX29830-62EA 27378.75 INR InStock
MFLX29830-62
Iron Reduction Cartridge for "Big Blue" housing
Filters Filter Cartridges
Reduce iron and metallic taste in water.
  • Designed with Permanganese filter media for iron reduction
Iron Reduction Cartridge provides an easy yet effective way to reduce iron in water. Reduces the metallic taste of water and helps eliminate stains on plumbing fixtures. Ideal for drinking water.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms