Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Masterflex® Console Digital Dispensing Drive with Remote Control for Micropump A Mount Gear Pumps, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Masterflex®
MFLX75211-35EA 0 INR InStock
MFLX75211-35
Masterflex® Console Digital Dispensing Drive with Remote Control for Micropump A Mount Gear Pumps, Avantor®
Pumps Pump Drives Gear Pump Drives
Easily operate this console drive from the front panel or use the optional foot switch (sold separately) for hands-free on/off control. The IP23-rated enclosure protects internal components from damage. Digital optical encoder feedback signal keeps speed control to ±0.3%.
What's included: the 115 VAC model has a 6-ft modular line cord with U.S. standard plug. The 230 VAC model has a 6-ft modular line cord with IEC 320/CEE 22 connector — please specify country to obtain correct plug when ordering.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms