Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Immersed Vertical Centrifugal Pumps, CPVC
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
70611-10 70611-20
MFLX70611-10EA 226000 INR InStock
MFLX70611-10 MFLX70611-20
Immersed Vertical Centrifugal Pumps, CPVC
Pumps Centrifugal Pumps
Immersed design eliminates need for seals and bearings—enables run dry operation.
  • Pump head located at the bottom of the pump minimizes cavitation
  • Compound impeller and vapor barrier separate fluid from motor
These immersed vertical centrifugal pumps are a sensible and economical choice for circulating, fume scrubbing, sump draining, agitation transfer, waste treatment, and wet processes. All pumps are constructed with a stainless steel (SS) shaft with sleeved CPVC or PVDF.
Delivery information: Includes polypropylene mounting plate.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms