Sanitary Centrifugal tube sets for viscosities up to 1200 cp
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
70603-02 70603-04
MFLX70603-02EA 509000 INR
MFLX70603-02 MFLX70603-04
Sanitary Centrifugal tube sets for viscosities up to 1200 cp
Pumps Centrifugal Pumps
Completely disassemble, clean, and re-assemble these stainless steel tube sets in 10 minutes or less!
  • Constructed of durable stainless steel (SS)
Centrifugal Tube Sets are ideal for low viscosity liquids such as wines, flavorings, perfumes, and sauces. Sets handle viscosities up to 1200 cp. Order a smaller 1/2-hp motor.