Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Parker High-Pressure Metal Three-way Ball Valves
Supplier: Parker
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX30527-80EA 26000 INR InStock
MFLX30527-80 MFLX30527-82 MFLX30527-60 MFLX30527-58 MFLX30527-88 MFLX30527-54 MFLX30527-56 MFLX30527-62 MFLX30527-84 MFLX30527-86
Parker High-Pressure Metal Three-way Ball Valves
Valves Ball Valves
Don’t worry about leaks.
  • Constructed of metal parts for durable applications
Ball valves are designed primarily for on/off service. The three-way valve model has three ports, and a combination of the ports can be either open or closed for various flow pattern capabilities. The handle of the valve can be turned to change the flow pattern or on/off service of the valve. Choose from stainless steel/PFA or brass/PFA valve options.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms