Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Hayward® Vacuum Breaker Vent Valves, PVC
Supplier: Hayward
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX06225-95EA 36942.26 INR InStock
MFLX06225-95
Hayward® Vacuum Breaker Vent Valves, PVC
Valves Air Regulation Valves
Eliminate problems when evacuating closed tanks or piping systems.
  • Constructed of durable PVC with a Viton® seal
Eliminates problems when evacuating closed tanks or piping systems. The one-way valve that is normally closed at positive pressure will allow drainage at a rate of 300 GPM. For higher drainage rates use additional vacuum breakers. The 3⁄4" NPT(F)socket adapts easily onto a 11⁄2" bulkhead fitting. The breaker is made of PVC with a Viton® seal.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms