Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Rechargeable Electric Backpack Sprayer
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX70605-05EA 123000 INR InStock
MFLX70605-05
Rechargeable Electric Backpack Sprayer
Spray Guns
Microprocessor pump control with adjustable spray tip – delicate mist, high volume, or long-range stream.
  • Tank capacity of 4 gallons
  • Comfortable ergonomic design with a full back pad and thickly padded shoulder harness.
  • Battery pack charges while in place or can be removed for remote charging.
Lightweight polyethylene tank is engineered to match the body's contour. Hand lever provides on-demand operation — depress to begin spraying and release to stop. Hand lever may also be locked in the open position to reduce operator fatigue during longer operation. Microprocessor regulates pump speed for consistent spraying at all four pump settings. High settings are great for a dense spray to cover large areas while low speeds are ideal to obtain a light mist for delicate work. A status lamp indicates when the power is on and blinks when the battery needs recharging. Battery life is 1000 cycles.
What's included: pump, tank with lid, 12 VDC battery, battery charger, 24'L spray wand, spray hose, inlet strainer, harness, and waist belt.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms