Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
316 Stainless Steel Investment Cast Close-Coupled Standard Centrifugal Pumps
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX73031-40EA 277933.09 INR InStock
MFLX73031-40 MFLX73031-20 MFLX73031-00 MFLX73031-35
316 Stainless Steel Investment Cast Close-Coupled Standard Centrifugal Pumps
Pumps Centrifugal Pumps
Open impellers handle clean fluids and fluids containing suspended solids.
  • Heavy-duty, stainless steel design—ideal for aggressive chemicals and high purity solutions
  • Rotate head in 90° increments for multiple mounting configurations
  • Viton® mechanical seal offers superior chemical resistance and accepts high inlet pressures
These close-coupled centrifugal pumps are constructed with a 316 stainless steel (SS) pump head, impeller, and shaft, along with Viton seals with carbon and ceramic faces. The open impeller design can handle clean fluids and fluids containing suspended solids.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms