Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Thermo Scientific™ Nalgene™ 180 Clear PVC Tubing
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
96700-20 96700-22 96700-23 96700-13
MFLX96700-20EA 90500 INR InStock
MFLX96700-20 MFLX96700-22 MFLX96700-23 MFLX96700-13
Thermo Scientific™ Nalgene™ 180 Clear PVC Tubing
Tubing Transfer Tubing
Resist to hardening and discoloration.
  • Global market leader in range of formulations for most extensive chemical compatibility
Thermo Scientific Nalgene 180 clear PVC tubing is ideal for applications in pharmaceutical, biotechnology, and general laboratory use. Resistant to hardening and discoloration, it offers good chemical resistance. The PVC resin meets USP Class VI and USDA requirements and may be sterilized either by autoclaving or with ethylene oxide. Usable from -26 to 160deg F (-32 to 71deg C).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms