Male Cam to Male NPT Threaded Adapters, Stainless Steel
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX34023-02EA 4786.67 INR
MFLX34023-02
Male Cam to Male NPT Threaded Adapters, Stainless Steel
Fittings and Connectors
Cam couplers provide quick disconnection from system.
  • All fittings have EPDM gaskets
  • Constructed of durable stainless steel
Cam couplers are ideal for providing a quick-disconnect solution to your pump line.