Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Flowline EchoSpan Ultrasonic Level Transmitters (2-Wire)
Supplier: Flowline
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX43260-04EA 191680.85 INR InStock
MFLX43260-04 MFLX43312-65 MFLX43260-00
Flowline EchoSpan Ultrasonic Level Transmitters (2-Wire)
Level Sensors
Setups fast with push-button calibration.
  • Offered in three measurement ranges up to 10 m, with 2" dia transducer
  • 6-segment LCD display indicates level in inch or centimeter values
  • 7.6 cm minimum beam width for applications with restricted space
  • PC/ABS enclosure rated NEMA 4X with rugged PVDF transducer
  • Fail-safe intelligence with diagnostic feedback for easy troubleshooting
EchoSpanâ„¢, a general-purpose two-wire ultrasonic transmitter, provides noncontact level measurement up to 32 ft or 10 m, and is ideally suited for challenging ultrapure, corrosive or waste liquids. Push-button calibrated, the transmitter is broadly selected for atmospheric bulk storage, day tank and waste sump applications. Media examples include wastewater and sodium hydroxide.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms