Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Kobold® Float Type Flow Switches, Brass
Supplier: Kobold
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX32481-45EA 60698.1 INR InStock
MFLX32481-45
Kobold® Float Type Flow Switches, Brass
Flow Switches
Small, compact design with low weight.
  • Constructed of durable brass
The device works on the well known float principle but without the use of the open conical measuring tube. Instead, the instruments use a cylindrical control tube with a conical groove along its axis. The flow switch can be used for continuous control of fluids and gases to ensure that excess or low flow cannot result in damage to the equipment. Applications can be found wherever it is necessary to control flow reliably.
FEATURES
- ± 4% Full scale accuracy
- Small, compact design, low weight
- Direct reading scales for water and air
- Large and small measuring ranges from 28:1 to 2.6:1 turndown
- Vertical connections for easy installation
- All media contacted parts available in brass or stainless steel construction
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms