Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Metric size stainless steel compression fittings
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX31528-12EA 6400 INR InStock
MFLX31528-12 MFLX31528-27
Metric size stainless steel compression fittings
Fittings and Connectors
Get a secure seal at higher pressures.
  • Constructed of durable stainless steel
Available in multiple configurations and sizes up to 12 mm. Stainless steel construction ensures very good chemical resistance and is suitable for high pressure (up to 10,000 psi / 689 bar) and high temperature (up to 1200 °F, 649 °C) applications.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms